missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-3-methyl-1-butanol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Specifications
Density | 0.926 |
Boiling Point | 173°C to 175°C |
Flash Point | 71°C (159°F) |
Refractive Index | 1.428 |
Quantity | 100 mL |
Beilstein | 1732882 |
Solubility Information | Soluble in water. |
Formula Weight | 118.18 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Methoxy-3-methyl-1-butanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'