missing translation for 'onlineSavingsMsg'
Learn More

3-(Methanesulfonyl)phenylboronic acid, 98%, ACROS Organics™

Brand:  Acros Organics 454240050

Additional Details : CAS Number : 373384-18-0 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15209926

  • 5880.34 NOK / 5g


Estimated Shipment
27-07-2020
Add to basket


Specifications

Chemical Identifiers

373384-18-0
200.02
3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
(3-methylsulfonylphenyl)boronic acid
C7H9BO4S
HZFFUMBZBGETES-UHFFFAOYSA-N
2773542
B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O

Specifications

3-(Methanesulfonyl)phenylboronic acid
102.0°C to 110.0°C
373384-18-0
C7H9BO4S
HZFFUMBZBGETES-UHFFFAOYSA-N
(3-methylsulfonylphenyl)boronic acid
2773542
98%
Authentic
5g
97.5% min. (HPLC)
3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O
200.02
200.02
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(Methanesulfonyl)phenylboronic acid, 98%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents