missing translation for 'onlineSavingsMsg'
Learn More

3-(Methanesulfonyl)phenylboronic acid, 98%, Thermo Scientific™

Catalog Number p-7077467
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
373384-18-0
C7H9BO4S
200.02
HZFFUMBZBGETES-UHFFFAOYSA-N
3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
2773542
(3-methylsulfonylphenyl)boronic acid
B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O
This item is not returnable. View return policy
373384-18-0
C7H9BO4S
200.02
HZFFUMBZBGETES-UHFFFAOYSA-N
3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
2773542
(3-methylsulfonylphenyl)boronic acid
B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O
Melting Point 102.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 200.02
Percent Purity 98%
Chemical Name or Material 3-(Methanesulfonyl)phenylboronic acid
compliance-icons
Product Identifier
  • 3-(Methanesulfonyl)phenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'