missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Maleimidopropionic acid, 95%, Thermo Scientific Chemicals
2088.10 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.
Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.
Notes
Moisture sensitive. Store in cool place.
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.
Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.
Notes
Moisture sensitive. Store in cool place.
Specifications
Melting Point | 108°C to 112°C |
Color | White |
Quantity | 1 g |
Beilstein | 1528952 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in methanol,dimethyl formamide,alcohol and dimethyl sulfoxide. Slightly soluble in water. |
Formula Weight | 169.13 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 3-Maleimidopropionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'