missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxybenzylamine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
3-Hydroxybenzylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxybenzylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water and ethanol.
Notes
Air sensitive. Incompatible with oxidizing agents.
3-Hydroxybenzylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water and ethanol.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | 170°C to 173°C |
Quantity | 250 mg |
UN Number | UN3259 |
Sensitivity | Air sensitive |
Solubility Information | Soluble in water and ethanol. |
Formula Weight | 123.15 |
Percent Purity | 97% |
Chemical Name or Material | 3-Hydroxybenzylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'