missing translation for 'onlineSavingsMsg'
Learn More

3'-Formyl(1,1'-biphenyl)-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3075505
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
222180-19-0
C14H10O3
226.231
MFCD04039068
QOCUSJPHQSKJJR-UHFFFAOYSA-N
3'-formyl-1,1'-biphenyl-3-carboxylic acid, 3-3-formylphenyl benzoic acid, 3'-formylbiphenyl-3-carboxylic acid, 3'-formyl 1,1'-biphenyl-3-carboxylic acid, 3'-formyl-biphenyl-3-carboxylic acid, amtda124, acmc-1cb16, 1,1'-biphenyl-3-carboxylicacid, 3'-formyl
2794721
3-(3-formylphenyl)benzoic acid
C1=CC(=CC(=C1)C=O)C2=CC(=CC=C2)C(=O)O
This item is not returnable. View return policy
222180-19-0
C14H10O3
226.231
MFCD04039068
QOCUSJPHQSKJJR-UHFFFAOYSA-N
3'-formyl-1,1'-biphenyl-3-carboxylic acid, 3-3-formylphenyl benzoic acid, 3'-formylbiphenyl-3-carboxylic acid, 3'-formyl 1,1'-biphenyl-3-carboxylic acid, 3'-formyl-biphenyl-3-carboxylic acid, amtda124, acmc-1cb16, 1,1'-biphenyl-3-carboxylicacid, 3'-formyl
2794721
3-(3-formylphenyl)benzoic acid
C1=CC(=CC(=C1)C=O)C2=CC(=CC=C2)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 3'-Formyl(1, 1'-biphenyl)-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'