missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7045025
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1711-07-5
C7H4ClFO
158.556
MFCD00000670
SYVNVEGIRVXRQH-UHFFFAOYSA-N
m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride
74376
3-fluorobenzoyl chloride
C1=CC(=CC(=C1)F)C(=O)Cl
This item is not returnable. View return policy
1711-07-5
C7H4ClFO
158.556
MFCD00000670
SYVNVEGIRVXRQH-UHFFFAOYSA-N
m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride
74376
3-fluorobenzoyl chloride
C1=CC(=CC(=C1)F)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -30°C
Density 1.32
Boiling Point 188°C to 189°C
Flash Point 82°C (179°F)
Odor Pungent
Refractive Index 1.5285
Quantity 5 g
UN Number UN3265
Beilstein 636610
Sensitivity Moisture sensitive
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'