missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Boronate esters are air- and chromatography-stable and suitable for spectroscopic study. The Suzuki-Miyaura cross-coupling reaction can be used with boronate esters.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Boronate esters are air- and chromatography-stable and suitable for spectroscopic study. The Suzuki-Miyaura cross-coupling reaction can be used with boronate esters.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
Quantity | 1 g |
Formula Weight | 307.17 |
Percent Purity | 95% |
Chemical Name or Material | 3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'