missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-formylbenzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7056146
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
871126-15-7
C7H6BFO3
167.93
MFCD10697421
GVHWLCYABLWGIR-UHFFFAOYSA-N
3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
53412038
(3-fluoro-2-formylphenyl)boronic acid
B(C1=C(C(=CC=C1)F)C=O)(O)O
This item is not returnable. View return policy
871126-15-7
C7H6BFO3
167.93
MFCD10697421
GVHWLCYABLWGIR-UHFFFAOYSA-N
3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
53412038
(3-fluoro-2-formylphenyl)boronic acid
B(C1=C(C(=CC=C1)F)C=O)(O)O

Undergoes unprecedented tautomeric rearrangement in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 127°C
Quantity 1 g
UN Number UN3261
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 167.93
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-formylbenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'