missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-1,2-dimethyl-6-nitrobenzene, 98%, Thermo Scientific™

Catalog Number p-7031470
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1736-87-4
C8H8FNO2
169.155
MFCD05663712
GLDMIZKOJPVEIV-UHFFFAOYSA-N
2,3-dimethyl-4-fluoronitrobenzene, 3-fluoro-6-nitro-1,2-dimethylbenzene, 3-fluoro-6-nitro-o-xylene, benzene, 1-fluoro-2,3-dimethyl-4-nitro, 3-fluoro-1,2-dimethyl-6-nitrobenzene, 4-fluoro-2,3-dimethyl nitrobenzene, intermediates-zcf02291, ksc534o2t, 2,3-dimethyl-1-fluoro-4-nitrobenzene, 4-fluoro-2,3-dimethyl-1-nitrobenzene
24721201
1-fluoro-2,3-dimethyl-4-nitrobenzene
CC1=C(C=CC(=C1C)F)[N+](=O)[O-]
This item is not returnable. View return policy
1736-87-4
C8H8FNO2
169.155
MFCD05663712
GLDMIZKOJPVEIV-UHFFFAOYSA-N
2,3-dimethyl-4-fluoronitrobenzene, 3-fluoro-6-nitro-1,2-dimethylbenzene, 3-fluoro-6-nitro-o-xylene, benzene, 1-fluoro-2,3-dimethyl-4-nitro, 3-fluoro-1,2-dimethyl-6-nitrobenzene, 4-fluoro-2,3-dimethyl nitrobenzene, intermediates-zcf02291, ksc534o2t, 2,3-dimethyl-1-fluoro-4-nitrobenzene, 4-fluoro-2,3-dimethyl-1-nitrobenzene
24721201
1-fluoro-2,3-dimethyl-4-nitrobenzene
CC1=C(C=CC(=C1C)F)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 39°C to 42°C
UN Number UN2811
Formula Weight 169.16
Percent Purity 98%
Quantity 250 mg
Odor Characteristic
Chemical Name or Material 3-Fluoro-1,2-dimethyl-6-nitrobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'