missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexylhydantoin, 95%, Thermo Scientific™

Catalog Number 15429508
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
67518-23-4
C9H14N2O2
182.223
MFCD12137796
SLCFSZHXCYYXLK-UHFFFAOYSA-N
3-cyclohexylhydantoine, 3-cyclohexyl-2,4-imidazolidinedione
12434609
3-cyclohexylimidazolidine-2,4-dione
C1CCC(CC1)N2C(=O)CNC2=O
This item is not returnable. View return policy
67518-23-4
C9H14N2O2
182.223
MFCD12137796
SLCFSZHXCYYXLK-UHFFFAOYSA-N
3-cyclohexylhydantoine, 3-cyclohexyl-2,4-imidazolidinedione
12434609
3-cyclohexylimidazolidine-2,4-dione
C1CCC(CC1)N2C(=O)CNC2=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 182.22
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Cyclohexylhydantoin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'