Learn More
3-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3-Chloro-2-fluorobenzeneboronic acid, Is used inorganic synthesis, catalysis, analytical chemistry and biological systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-2-fluorobenzeneboronic acid, Is used inorganic synthesis, catalysis, analytical chemistry and biological systems.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
3-Chloro-2-fluorobenzeneboronic acid, Is used inorganic synthesis, catalysis, analytical chemistry and biological systems.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 240°C to 242°C |
Quantity | 1 g |
Solubility Information | Insoluble in water. Soluble in organic solvents. |
Formula Weight | 174.37 |
Percent Purity | 97% |
Chemical Name or Material | 3-Chloro-2-fluorobenzeneboronic acid |