missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7027494
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
186517-45-3
C8H2Cl2F4O
260.997
MFCD01631344
QRJJXSXDJDIRPI-UHFFFAOYSA-N
3-chloro-2-fluoro-6-trifluoromethyl benzoyl chloride, 3-chloro-2-fluoro-6-trifluoromethyl benzoylchloride, benzoyl chloride,2-chloro-3-fluoro-6-trifluoromethyl, acmc-1c02f, 3-chloro-2-fluoro-6-trifluoromethyl-benzoyl chloride, 3-chloranyl-2-fluoranyl-6-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-chloro-2-fluoro-6-trifluoromethyl
2773784
3-chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C(=C1C(F)(F)F)C(=O)Cl)F)Cl
This item is not returnable. View return policy
186517-45-3
C8H2Cl2F4O
260.997
MFCD01631344
QRJJXSXDJDIRPI-UHFFFAOYSA-N
3-chloro-2-fluoro-6-trifluoromethyl benzoyl chloride, 3-chloro-2-fluoro-6-trifluoromethyl benzoylchloride, benzoyl chloride,2-chloro-3-fluoro-6-trifluoromethyl, acmc-1c02f, 3-chloro-2-fluoro-6-trifluoromethyl-benzoyl chloride, 3-chloranyl-2-fluoranyl-6-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-chloro-2-fluoro-6-trifluoromethyl
2773784
3-chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C(=C1C(F)(F)F)C(=O)Cl)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.475
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 261
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'