missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,6-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7027269
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
225104-76-7
C7H3ClF2O2
192.55
MFCD01631322
APQTVWJSXBBNEI-UHFFFAOYSA-N
2773534
3-chloro-2,6-difluorobenzoic acid
OC(=O)C1=C(F)C=CC(Cl)=C1F
This item is not returnable. View return policy
225104-76-7
C7H3ClF2O2
192.55
MFCD01631322
APQTVWJSXBBNEI-UHFFFAOYSA-N
2773534
3-chloro-2,6-difluorobenzoic acid
OC(=O)C1=C(F)C=CC(Cl)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 126°C to 128°C
Quantity 1 g
Synonym 2,6-difluoro-3-chlorobenzoic acid, 2,6-difluouo-3-chlorobenzoic acid, benzoic acid, 3-chloro-2,6-difluoro, 3-chloro-2,6-difluoro-benzoicacid, 3-chloro-2,6-difluoro-benzoic acid, pubchem8531, acmc-1cqaj, rarechem al be 0491, benzoic acid,3-chloro-2,6-difluoro
Formula Weight 192.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,6-difluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'