missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,6-difluorobenzamide, 97+%, Thermo Scientific™

Catalog Number p-7027366
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
261762-41-8
C7H4ClF2NO
191.562
MFCD01631386
TUTOELHQVJTZTO-UHFFFAOYSA-N
acmc-1cfko, benzamide,3-chloro-2,6-difluoro
2773517
3-chloro-2,6-difluorobenzamide
C1=CC(=C(C(=C1F)C(=O)N)F)Cl
This item is not returnable. View return policy
261762-41-8
C7H4ClF2NO
191.562
MFCD01631386
TUTOELHQVJTZTO-UHFFFAOYSA-N
acmc-1cfko, benzamide,3-chloro-2,6-difluoro
2773517
3-chloro-2,6-difluorobenzamide
C1=CC(=C(C(=C1F)C(=O)N)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 133°C to 136°C
Quantity 1 g
Formula Weight 191.56
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-2,6-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'