missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamoyl-5-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7034961
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871332-66-0
C7H7BFNO3
182.945
MFCD08056357
FTMWTTRIUYYHDE-UHFFFAOYSA-N
3-carbamoyl-5-fluorophenyl boronic acid, 3-borono-5-fluorobenzamide, 3-aminocarbonyl-5-fluorobenzeneboronic acid, acmc-209qgw, 3-carbamoyl-5-fluorobenzeneboronic acid, 3-aminocarbonyl-5-fluoranyl-phenyl boronic acid, boronic acid,b-3-aminocarbonyl-5-fluorophenyl
44717588
(3-carbamoyl-5-fluorophenyl)boronic acid
B(C1=CC(=CC(=C1)F)C(=O)N)(O)O
This item is not returnable. View return policy
871332-66-0
C7H7BFNO3
182.945
MFCD08056357
FTMWTTRIUYYHDE-UHFFFAOYSA-N
3-carbamoyl-5-fluorophenyl boronic acid, 3-borono-5-fluorobenzamide, 3-aminocarbonyl-5-fluorobenzeneboronic acid, acmc-209qgw, 3-carbamoyl-5-fluorobenzeneboronic acid, 3-aminocarbonyl-5-fluoranyl-phenyl boronic acid, boronic acid,b-3-aminocarbonyl-5-fluorophenyl
44717588
(3-carbamoyl-5-fluorophenyl)boronic acid
B(C1=CC(=CC(=C1)F)C(=O)N)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 280°C to 286°C
Quantity 250 mg
Formula Weight 182.95
Percent Purity 98%
Chemical Name or Material 3-Carbamoyl-5-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'