Learn More
3-Bromobiphenyl, 99%, Thermo Scientific Chemicals
Catalog Number p-7025973
Click to view available options
Quantity:
1 g
5 g
25 g
2113-57-7
C12H9Br
233.11
MFCD00000082
USYQKCQEVBFJRP-UHFFFAOYSA-N
3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
16449
1-bromo-3-phenylbenzene
BrC1=CC=CC(=C1)C1=CC=CC=C1
2113-57-7
C12H9Br
233.11
MFCD00000082
USYQKCQEVBFJRP-UHFFFAOYSA-N
3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
16449
1-bromo-3-phenylbenzene
BrC1=CC=CC(=C1)C1=CC=CC=C1
3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.397
Boiling Point 301°C
Flash Point >110°C (230°F)
Refractive Index 1.641
Quantity 1 g
Beilstein 2043191
Solubility Information Immiscible with water.
Formula Weight 233.12
Percent Purity 99%
Chemical Name or Material 3-Bromobiphenyl