Learn More
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
Catalog Number p-72907
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
823-78-9
C7H6Br2
249.93
MFCD00000176
ZPCJPJQUVRIILS-UHFFFAOYSA-N
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
69979
1-bromo-3-(bromomethyl)benzene
C1=CC(=CC(=C1)Br)CBr
823-78-9
C7H6Br2
249.93
MFCD00000176
ZPCJPJQUVRIILS-UHFFFAOYSA-N
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
69979
1-bromo-3-(bromomethyl)benzene
C1=CC(=CC(=C1)Br)CBr
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 37.0°C to 42.0°C
Color White
Boiling Point 130.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2Br
Quantity 5 g
Beilstein 05, 308
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 249.93
Percent Purity 95%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 3-Bromobenzyl bromide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.