Learn More
3-Bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals
Catalog Number p-4258798
Click to view available options
Quantity:
5 g
25 g
73583-39-8
C5H3BrClN
192.44
BELDOPUBSLPBCQ-UHFFFAOYSA-N
3-chloro-5-bromopyridine, 3-bromo-5-chloro-pyridine, pyridine, 3-bromo-5-chloro, 5-bromo-3-chloropyridine, pubchem7700, 3-brom-5-chlorpyridin, acmc-209orf, #, ksc377a4j, 3-bromo-5-chloropyridine
606256
3-bromo-5-chloropyridine
C1=C(C=NC=C1Br)Cl
73583-39-8
C5H3BrClN
192.44
BELDOPUBSLPBCQ-UHFFFAOYSA-N
3-chloro-5-bromopyridine, 3-bromo-5-chloro-pyridine, pyridine, 3-bromo-5-chloro, 5-bromo-3-chloropyridine, pubchem7700, 3-brom-5-chlorpyridin, acmc-209orf, #, ksc377a4j, 3-bromo-5-chloropyridine
606256
3-bromo-5-chloropyridine
C1=C(C=NC=C1Br)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 78.7°C to 80.1°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 192.44
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chloropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromo-5-chloropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.