missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybiphenyl, 99%, Thermo Scientific™

Catalog Number p-7033461
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
74447-73-7
C13H11BrO
263.13
MFCD00079720
QJGJVVIDRMVQMR-UHFFFAOYSA-N
3-bromo-4-methoxybiphenyl, 3-bromo-4-methoxy-1,1'-biphenyl, 2-bromo-1-methoxy-4-phenyl-menzene, acmc-20an8p, 2-bromo-4-phenylanisole, maybridge3_000975, 3-bromo-4-methoxy-biphenyl, 2-methoxy-5-phenyl-phenylbromide, 2-bromo-1-methoxy-4-phenyl-benzene
262267
2-bromo-1-methoxy-4-phenylbenzene
COC1=CC=C(C=C1Br)C1=CC=CC=C1
This item is not returnable. View return policy
74447-73-7
C13H11BrO
263.13
MFCD00079720
QJGJVVIDRMVQMR-UHFFFAOYSA-N
3-bromo-4-methoxybiphenyl, 3-bromo-4-methoxy-1,1'-biphenyl, 2-bromo-1-methoxy-4-phenyl-menzene, acmc-20an8p, 2-bromo-4-phenylanisole, maybridge3_000975, 3-bromo-4-methoxy-biphenyl, 2-methoxy-5-phenyl-phenylbromide, 2-bromo-1-methoxy-4-phenyl-benzene
262267
2-bromo-1-methoxy-4-phenylbenzene
COC1=CC=C(C=C1Br)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 263.14
Percent Purity 99%
Chemical Name or Material 3-Bromo-4-methoxybiphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'