missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloropyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-2699890
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
52200-48-3
C5H3BrClN
192.44
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br
This item is not returnable. View return policy
52200-48-3
C5H3BrClN
192.44
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 57.9°C to 58.3°C
Color White to Yellow
Boiling Point 97°C (1.3 kPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 192.44
Percent Purity 98%
Physical Form Crystalline Powder, Crystals, Flakes or Solid
Chemical Name or Material 3-Bromo-2-chloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-2-chloropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'