Learn More
Alfa Aesar™ 3-Bromo-2-chloro-6-fluorotoluene, 97%
Brand:  Alfa Aesar™ H63276.06
Additional Details : CAS Number : 203302-92-5 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15429898
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
203302-92-5
223.469
VHQALVHKNXDJRY-UHFFFAOYSA-N
2736227
CC1=C(C=CC(=C1Cl)Br)F
C7H5BrClF
MFCD00236591
3-bromo-2-chloro-6-fluorotoluene, 2-chloro-3-bromo-6-fluorotoluene, 3-bromo-2-chlor-6-fluorotoluene, benzene, 1-bromo-2-chloro-4-fluoro-3-methyl, castest, pubchem4514, 1-bromo-2-chloro-4-fluoro-3-methyl-benzene, acmc-1ce0p, 3-bromo-2-chloro-6-fiuorotoluene, 3-bromo-2-chloro-6-fluoro-toluene
1-bromo-2-chloro-4-fluoro-3-methylbenzene
Specifications
3-Bromo-2-chloro-6-fluorotoluene
203302-92-5
MFCD00236591
VHQALVHKNXDJRY-UHFFFAOYSA-N
1-bromo-2-chloro-4-fluoro-3-methylbenzene
2736227
Solid
5g
C7H5BrClF
3-bromo-2-chloro-6-fluorotoluene, 2-chloro-3-bromo-6-fluorotoluene, 3-bromo-2-chlor-6-fluorotoluene, benzene, 1-bromo-2-chloro-4-fluoro-3-methyl, castest, pubchem4514, 1-bromo-2-chloro-4-fluoro-3-methyl-benzene, acmc-1ce0p, 3-bromo-2-chloro-6-fiuorotoluene, 3-bromo-2-chloro-6-fluoro-toluene
CC1=C(C=CC(=C1Cl)Br)F
223.469
223.47
97%