Learn More
3-Amino-4-ethyl-1H-pyrazole, 98%, Thermo Scientific Chemicals
Catalog Number p-7054788
Click to view available options
Quantity:
250 mg
1 g
5 g
43024-15-3
C5H9N3
111.15
MFCD06797570
RDCODVKTTJWFAR-UHFFFAOYSA-N
4-ethyl-1h-pyrazol-3-amine, 3-amino-4-ethylpyrazole, 3-amino-4-ethyl-1h-pyrazole, 4-ethyl-2h-pyrazol-3-amine, 3-amino-4-ethyl pyrazole, 4-ethyl-1h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 4-ethyl
11789157
4-ethyl-1H-pyrazol-5-amine
CCC1=C(N)NN=C1
43024-15-3
C5H9N3
111.15
MFCD06797570
RDCODVKTTJWFAR-UHFFFAOYSA-N
4-ethyl-1h-pyrazol-3-amine, 3-amino-4-ethylpyrazole, 3-amino-4-ethyl-1h-pyrazole, 4-ethyl-2h-pyrazol-3-amine, 3-amino-4-ethyl pyrazole, 4-ethyl-1h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 4-ethyl
11789157
4-ethyl-1H-pyrazol-5-amine
CCC1=C(N)NN=C1
3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 111.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-4-ethyl-1H-pyrazole