Learn More
Alfa Aesar™ 3-Amino-2-chloro-4-methylpyridine, 97%
Brand: Alfa Aesar™ H26058.06
Additional Details : CAS Number : 133627-45-9 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
133627-45-9 | |
142.586 | |
UOBCYTOUXLAABU-UHFFFAOYSA-N | |
2756387 | |
CC1=C(C(=NC=C1)Cl)N |
C6H7ClN2 | |
MFCD00673152 | |
3-amino-2-chloro-4-methylpyridine, 2-chloro-3-amino-4-methylpyridine, 3-amino-2-chloro-4-picoline, 2-chloro-3-amino-4-methyl pyridine, 2-chloro-4-methyl-pyridin-3-ylamine, 2-chloro-3-amino-4-picoline, 2-chloro-4-methyl-3-pyridinamine, 3-pyridinamine, 2-chloro-4-methyl, 2-chloro-4-methyl-3-pyridylamine, 3-amino-2-chloro-4-methyl pyridine | |
2-chloro-4-methylpyridin-3-amine |
Specifications
3-Amino-2-chloro-4-methylpyridine | |
5g | |
C6H7ClN2 | |
3-amino-2-chloro-4-methylpyridine, 2-chloro-3-amino-4-methylpyridine, 3-amino-2-chloro-4-picoline, 2-chloro-3-amino-4-methyl pyridine, 2-chloro-4-methyl-pyridin-3-ylamine, 2-chloro-3-amino-4-picoline, 2-chloro-4-methyl-3-pyridinamine, 3-pyridinamine, 2-chloro-4-methyl, 2-chloro-4-methyl-3-pyridylamine, 3-amino-2-chloro-4-methyl pyridine | |
CC1=C(C(=NC=C1)Cl)N | |
142.586 | |
142.59 |
64°C to 68°C | |
133627-45-9 | |
MFCD00673152 | |
UOBCYTOUXLAABU-UHFFFAOYSA-N | |
2-chloro-4-methylpyridin-3-amine | |
2756387 | |
97% |