missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylpyridine adenine dinucleotide, 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Infrared Spectrum | Authentic |
Assay Percent Range | 89% min. (HPLC) |
Quantity | 250 mg |
IUPAC Name | 3-acetyl-1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium |
Formula Weight | 662.45 |
Percent Purity | 90% |
Chemical Name or Material | 3-Acetylpyridine adenine dinucleotide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'