missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7081385
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
60481-36-9
C8H13ClN2
172.656
MFCD00052269
RSBQANBNDXZFIY-UHFFFAOYSA-N
3,5-dimethylphenylhydrazine hydrochloride, 3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenylhydrazinehydrochloride, 3,5-dimethyl phenylhydrazine hydrochloride, 3,5-dimethylphenylhydrazine, hydrazine, 3,5-dimethylphenyl-, monohydrochloride, pubchem6407, ksc491e7r, 1-3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenyl hydrazine-hydrogen chloride 1/1
12651341
(3,5-dimethylphenyl)hydrazine;hydrochloride
CC1=CC(=CC(=C1)NN)C.Cl
This item is not returnable. View return policy
60481-36-9
C8H13ClN2
172.656
MFCD00052269
RSBQANBNDXZFIY-UHFFFAOYSA-N
3,5-dimethylphenylhydrazine hydrochloride, 3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenylhydrazinehydrochloride, 3,5-dimethyl phenylhydrazine hydrochloride, 3,5-dimethylphenylhydrazine, hydrazine, 3,5-dimethylphenyl-, monohydrochloride, pubchem6407, ksc491e7r, 1-3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenyl hydrazine-hydrogen chloride 1/1
12651341
(3,5-dimethylphenyl)hydrazine;hydrochloride
CC1=CC(=CC(=C1)NN)C.Cl

3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 180°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylphenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'