missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methoxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15431269
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
650140-84-4
C6H5Br2NO2
282.919
MFCD13185524
KAYQEYHOGFRTAL-UHFFFAOYSA-N
3,5-dibromo-4-methoxypyridine 1-oxide, 3,5-dibromo-4-methoxypyridine-n-oxide, 3,5-dibromo-4-methoxypyridin-1-ium-1-olate, pubchem6614
2784723
3,5-dibromo-4-methoxy-1-oxidopyridin-1-ium
COC1=C(C=[N+](C=C1Br)[O-])Br
This item is not returnable. View return policy
650140-84-4
C6H5Br2NO2
282.919
MFCD13185524
KAYQEYHOGFRTAL-UHFFFAOYSA-N
3,5-dibromo-4-methoxypyridine 1-oxide, 3,5-dibromo-4-methoxypyridine-n-oxide, 3,5-dibromo-4-methoxypyridin-1-ium-1-olate, pubchem6614
2784723
3,5-dibromo-4-methoxy-1-oxidopyridin-1-ium
COC1=C(C=[N+](C=C1Br)[O-])Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 266.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3,5-Dibromo-4-methoxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'