missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Catalog Number p-7033912
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1036991-25-9
C17H24BNO4
317.192
MFCD09266172
PKONGXZPUDFPPH-UHFFFAOYSA-N
morpholino 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 3-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 3-4-morpholinylcarbonyl phenylboronic acid pinacol ester, 3-morpholine-4-carbonyl phenyl boronic acid pinacol ester, amtb027, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine
46739724
morpholin-4-yl-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)N3CCOCC3
This item is not returnable. View return policy
1036991-25-9
C17H24BNO4
317.192
MFCD09266172
PKONGXZPUDFPPH-UHFFFAOYSA-N
morpholino 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 3-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 3-4-morpholinylcarbonyl phenylboronic acid pinacol ester, 3-morpholine-4-carbonyl phenyl boronic acid pinacol ester, amtb027, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine
46739724
morpholin-4-yl-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)N3CCOCC3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 317.19
Chemical Name or Material 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'