Learn More
3,4-Dimethoxyaniline, 98+%, Thermo Scientific Chemicals
Catalog Number p-7021275
Click to view available options
Quantity:
10 g
50 g
250 g
6315-89-5
C8H11NO2
153.181
MFCD00008394
LGDHZCLREKIGKJ-UHFFFAOYSA-N
4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
22770
3,4-dimethoxyaniline
COC1=C(C=C(C=C1)N)OC
6315-89-5
C8H11NO2
153.181
MFCD00008394
LGDHZCLREKIGKJ-UHFFFAOYSA-N
4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
22770
3,4-dimethoxyaniline
COC1=C(C=C(C=C1)N)OC
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
Solubility
Slightly soluble in water
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 86°C to 89°C
Boiling Point 174°C to 176°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 10 g
UN Number UN2811
Beilstein 743399
Solubility Information Slightly soluble in water
Formula Weight 153.18
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxyaniline