missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65294.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429546

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD00684199

Specifications

3,4-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
MFCD00684199
Solid
1g
311.38
95%
Videos
SDS Documents