Learn More
3,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.
Solubility
Soluble in water(6.3g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.
Solubility
Soluble in water(6.3g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | 152°C to 157°C |
Quantity | 5 g |
Beilstein | 774381 |
Sensitivity | Air Sensitive |
Merck Index | 14,7893 |
Solubility Information | Soluble in water(6.3g/L). |
Formula Weight | 138.12 |
Percent Purity | 98% |
Chemical Name or Material | 3,4-Dihydroxybenzaldehyde |