missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoroaniline, 98+%, Thermo Scientific™

Catalog Number 11407856
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3863-11-4
MFCD00007761
This item is not returnable. View return policy
3863-11-4
MFCD00007761

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 20°C to 22°C
Density 1.302
Boiling Point 196°C to 198°C
Flash Point 85°C (185°F)
Odor Characteristic
Molecular Formula C4H10N2O2
Refractive Index 1.513
Quantity 5 g
UN Number UN2941
Beilstein 971235
Synonym 4-hydroxybutyric acid hydrazide, 4-hydroxybutanohydrazide, butanoic acid, 4-hydroxy-, hydrazide, 4-???butan-1-ol, acmc-1cts6, 4-hydroxybutanoyl hydrazine, 4-hydroxybutanoic acid hydrazide, butanoic acid,4-hydroxy-, hydrazide
InChI Key WOXQFPPKZBOSTN-UHFFFAOYSA-N
SMILES C(CC(=O)NN)CO
IUPAC Name 4-hydroxybutanehydrazide
Molecular Weight (g/mol) 118.14
PubChem CID 220659
Formula Weight 129.11
Percent Purity ≥98%
Chemical Name or Material 3,4-Difluoroaniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'