missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals

Catalog Number 15460806
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

3,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store under inert gas. Store away fro
TRUSTED_SUSTAINABILITY
CAS 61259-69-6
Molecular Formula C7H5Cl3Mg
MDL Number MFCD01311472
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 3,4-dichlorobenzylmagnesium chloride, chloro 3,4-dichlorophenyl methyl magnesium, jtpvddbuxvullo-uhfffaoysa-m, magnesium, chloro 3,4-dichlorophenyl methyl, magnesium,1,2-dichloro-4-methanidylbenzene,chloride, 3,4-dichlorobenzylmagnesium chloride, 0.25m in 2-methf, 3,4-dichlorobenzylmagnesium chloride 0.25 m in diethyl ether
Solubility Information Reacts with water.
InChI Key JTPVDDBUXVULLO-UHFFFAOYSA-M
SMILES Cl[Mg]CC1=CC(Cl)=C(Cl)C=C1
IUPAC Name magnesium;1,2-dichloro-4-methanidylbenzene;chloride
Molecular Weight (g/mol) 219.77
PubChem CID 24722679
Formula Weight 219.78
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 3,4-Dichlorobenzylmagnesium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'