Learn More
3,4,5-Trimethoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog Number 10018039
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 133095-91-7, 109-99-9
CAS Min % 11.09, 83.37
CAS Max % 16.63, 88.91
Color Brown to Yellow
Density 0.9790g/mL
Flash Point −20°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C9H11BrMgO3
Linear Formula (OCH3)3C6H2MgBr
Quantity 100 mL
Specific Gravity 0.979
Synonym 3,4,5-trimethoxyphenylmagnesium bromide, 3,4,5-trimethoxyphenylmagnesium bromide solution, ywbltwdgwzpylj-uhfffaoysa-m, 3,4,5-trimethoxyphenylmagnesiumbromide, 3,4,5-trimethoxyphenyl magnesium bromide, 3,4,5-trimethoxyphenylmagnesium bromide, 0.25 m in thf, 3,4,5-trimethoxyphenylmagnesium bromide 0.25 m in tetrahydrofuran, 3,4,5-trimethoxyphenylmagnesium bromide solution, 0.5 m in thf, 3,4,5-trimethoxyphenylmagnesium bromide solution 0.5m in tetrahydrofuran, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key RXSCVXDQDANNME-UHFFFAOYSA-M
SMILES COC1=C(C(=C[C-]=C1)OC)OC.[Mg+2].[Br-]
IUPAC Name magnesium;1,2,3-trimethoxybenzene-5-ide;bromide
Molecular Weight (g/mol) 271.39
PubChem CID 11140221
Formula Weight 271.39
Physical Form Solution
Chemical Name or Material 3, 4, 5-Trimethoxyphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,4,5-Trimethoxyphenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.