missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Chlorophenyl)-2-thiohydantoin, 95%

990.00 NOK - 11880.00 NOK

Chemical Identifiers

MDL Number MFCD03070245
Synonym 3-(2-Chlorophenyl)-2-thioxo-4-imidazolidinone
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15439538 SDS View Documents Alfa Aesar™
H63061.MD
250mg 990.00 NOK
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-09-2019
Add to basket
 
15419538 SDS View Documents Alfa Aesar™
H63061.03
1g 2980.00 NOK
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-09-2019
Add to basket
 
15429538 SDS View Documents Alfa Aesar™
H63061.06
5g 11880.00 NOK
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-09-2019
Add to basket
 
Specifications

Chemical Identifiers

MFCD03070245
3-(2-Chlorophenyl)-2-thioxo-4-imidazolidinone

Specifications

3-(2-Chlorophenyl)-2-thiohydantoin
MFCD03070245
226.68
95%
250mg
3-(2-Chlorophenyl)-2-thioxo-4-imidazolidinone
Solid