Learn More
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™
Catalog Number p-4391299
Click to view available options
Quantity:
10 g
50 g
100 g
3919-76-4
C11H7Cl2NO3
272.08
MFCD00051658
WQXUUMUOERZZAE-UHFFFAOYSA-N
3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2',6'-dichlorophenyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-2,6-dichlorophenyl-5-methyl-4-isoxazolecarboxylic acid, pubchem8684, dicloxacillin impurity d, acmc-1cmb3, pyridinium hydroxy propyl sulphobetaine; pps-oh, 3-2,6-dichlorophenyl-5-methylisoxazol-4-carboxylic acid
77521
3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
CC1=C(C(O)=O)C(=NO1)C1=C(Cl)C=CC=C1Cl
3919-76-4
C11H7Cl2NO3
272.08
MFCD00051658
WQXUUMUOERZZAE-UHFFFAOYSA-N
3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2',6'-dichlorophenyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-2,6-dichlorophenyl-5-methyl-4-isoxazolecarboxylic acid, pubchem8684, dicloxacillin impurity d, acmc-1cmb3, pyridinium hydroxy propyl sulphobetaine; pps-oh, 3-2,6-dichlorophenyl-5-methylisoxazol-4-carboxylic acid
77521
3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
CC1=C(C(O)=O)C(=NO1)C1=C(Cl)C=CC=C1Cl
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 272.09
Percent Purity 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid