missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7034838
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850567-38-3
C8H9BClN5O3
269.452
MFCD06659888
PWSGJXZLSYWEAJ-UHFFFAOYSA-N
3-1h-tetrazol-5-yl carbamoyl phenyl boronic acid hydrochloride, 3-1h-tetrazol-5-ylcarbamoyl benzeneboronic acid hydrochloride, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid, hcl, 3-1h-tetrazol-5-yl-carbamoyl benzeneboronic acid, hcl, 3-1h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hcl, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hydrochloride, 3-1h-tetrazol-5-ylaminocarbonyl phenylboronic acid hydrochloride, 3-2h-tetrazol-5-yl carbamoyl phenyl boronic acid-hydrogen chloride 1/1, 3-2h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride
44119544
[3-(2H-tetrazol-5-ylcarbamoyl)phenyl]boronic acid;hydrochloride
B(C1=CC(=CC=C1)C(=O)NC2=NNN=N2)(O)O.Cl
This item is not returnable. View return policy
850567-38-3
C8H9BClN5O3
269.452
MFCD06659888
PWSGJXZLSYWEAJ-UHFFFAOYSA-N
3-1h-tetrazol-5-yl carbamoyl phenyl boronic acid hydrochloride, 3-1h-tetrazol-5-ylcarbamoyl benzeneboronic acid hydrochloride, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid, hcl, 3-1h-tetrazol-5-yl-carbamoyl benzeneboronic acid, hcl, 3-1h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hcl, 3-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hydrochloride, 3-1h-tetrazol-5-ylaminocarbonyl phenylboronic acid hydrochloride, 3-2h-tetrazol-5-yl carbamoyl phenyl boronic acid-hydrogen chloride 1/1, 3-2h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride
44119544
[3-(2H-tetrazol-5-ylcarbamoyl)phenyl]boronic acid;hydrochloride
B(C1=CC(=CC=C1)C(=O)NC2=NNN=N2)(O)O.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 250 mg
Formula Weight 269.45
Percent Purity 97%
Chemical Name or Material 3-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'