missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-yl)propanoic acid, ≥97%, Thermo Scientific™

Catalog Number p-3110325
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
18999-45-6
C6H8N2O2
140.142
MFCD02731119
VSFNAZLYGOOSEY-UHFFFAOYSA-N
3-1h-imidazol-1-yl propanoic acid, 3-imidazol-1-yl-propionic acid, 1h-imidazole-1-propanoic acid, 3-1-imidazolyl propanoic acid, chembl20724, 3-imidazolylpropanoic acid, 3-imidazol-1-yl-propionicacid, 3-1-imidazolyl propionic acid, 3 1h-imidazol-1-yl propanoic acid
2794718
3-imidazol-1-ylpropanoic acid
C1=CN(C=N1)CCC(=O)O
This item is not returnable. View return policy
18999-45-6
C6H8N2O2
140.142
MFCD02731119
VSFNAZLYGOOSEY-UHFFFAOYSA-N
3-1h-imidazol-1-yl propanoic acid, 3-imidazol-1-yl-propionic acid, 1h-imidazole-1-propanoic acid, 3-1-imidazolyl propanoic acid, chembl20724, 3-imidazolylpropanoic acid, 3-imidazol-1-yl-propionicacid, 3-1-imidazolyl propionic acid, 3 1h-imidazol-1-yl propanoic acid
2794718
3-imidazol-1-ylpropanoic acid
C1=CN(C=N1)CCC(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 3-(1H-imidazol-1-yl)propanoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'