missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 906352-85-0 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C11H10N2O2 |
MDL Number | MFCD09065006 |
Quantity | 1 g |
Synonym | 3-1-methyl-1h-pyrazol-3-yl benzoic acid, 3-1-methylpyrazol-3-yl benzoic acid, benzoicacid,3-1-methyl-1h-pyrazol-3-yl, 3-3-carboxyphenyl-1-methyl-1h-pyrazole, benzoicacid, 3-1-methyl-1h-pyrazol-3-yl, benzoic acid, 3-1-methyl-1h-pyrazol-3-yl |
InChI Key | NQPMBEUYTBLTAG-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'