missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-1-methyl-1h-pyrazol-3-yl benzenesulfonyl chloride, benzenesulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl benzenesulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl benzenesulphonyl chloride, 3-1-methyl-1h-pyrazol-3-yl benzene-1-sulfonyl chloride, chloro 3-1-methylpyrazol-3-yl phenyl sulfone, 3-1-methyl-1h-pyrazol-3-yl phenylsulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl-benzenesulfonyl chloride, benzenesulfonyl chloride,3-1-methyl-1h-pyrazol-3-yl |
Formula Weight | 256.71 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'