Learn More
Cytiva 2CHR Filter Paper
Brand:  Cytiva 3002-9159
5918.50 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.02000kg
Product Code. 15638298
  • 6230.00 NOK / Each
Estimated Shipment: 09-09-2024
Specifications
10 x 150 mm
2 CHR