missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™

Catalog Number p-7028986
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
238403-52-6
C8H6BrF3S
271.095
MFCD00236345
ZSBCQBRVNBLWAW-UHFFFAOYSA-N
2-trifluoromethylthio benzyl bromide, 2-trifluoromethylthio benzylbromide, 1-bromomethyl-2-trifluoromethylsulfanyl-benzene, 1-bromomethyl-2-trifluoromethylsulfanyl benzene, 1-bromomethyl-2-trifluoromethyl sulfanyl benzene, acmc-1ccq6, 2-trifluoromethylthiobenzylbromide, 2-trifluoromethyl thio benzyl bromide, 2-bromomethyl phenylthio trifluoromethane, 2-bromomethyl phenyl trifluoromethyl sulphide
2777876
1-(bromomethyl)-2-(trifluoromethylsulfanyl)benzene
C1=CC=C(C(=C1)CBr)SC(F)(F)F
This item is not returnable. View return policy
238403-52-6
C8H6BrF3S
271.095
MFCD00236345
ZSBCQBRVNBLWAW-UHFFFAOYSA-N
2-trifluoromethylthio benzyl bromide, 2-trifluoromethylthio benzylbromide, 1-bromomethyl-2-trifluoromethylsulfanyl-benzene, 1-bromomethyl-2-trifluoromethylsulfanyl benzene, 1-bromomethyl-2-trifluoromethyl sulfanyl benzene, acmc-1ccq6, 2-trifluoromethylthiobenzylbromide, 2-trifluoromethyl thio benzyl bromide, 2-bromomethyl phenylthio trifluoromethane, 2-bromomethyl phenyl trifluoromethyl sulphide
2777876
1-(bromomethyl)-2-(trifluoromethylsulfanyl)benzene
C1=CC=C(C(=C1)CBr)SC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 71°C (1 mmHg)
Refractive Index 1.5295
Quantity 1 g
UN Number UN1760
Formula Weight 271.1
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethylthio)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'