Learn More
2'-(Trifluoromethoxy)acetophenone, 98+%, Thermo Scientific™
Catalog Number 11389935
Click to view available options
Quantity:
1 g
5 g
220227-93-0
C9H7F3O2
204.148
MFCD00236320
XTXSTESGCXKUIH-UHFFFAOYSA-N
2'-trifluoromethoxy acetophenone, 1-2-trifluoromethoxy phenyl ethanone, 1-2-trifluoromethoxy phenyl ethan-1-one, 1-acetyl-2-trifluoromethoxy benzene, pubchem4295, 2-trifluoromethoxy acetophenone98, 1-2-trifluoromethoxyphenyl ethanone, 2'-trifluoromethoxy acetophenone98
2777176
1-[2-(trifluoromethoxy)phenyl]ethanone
CC(=O)C1=CC=CC=C1OC(F)(F)F
220227-93-0
C9H7F3O2
204.148
MFCD00236320
XTXSTESGCXKUIH-UHFFFAOYSA-N
2'-trifluoromethoxy acetophenone, 1-2-trifluoromethoxy phenyl ethanone, 1-2-trifluoromethoxy phenyl ethan-1-one, 1-acetyl-2-trifluoromethoxy benzene, pubchem4295, 2-trifluoromethoxy acetophenone98, 1-2-trifluoromethoxyphenyl ethanone, 2'-trifluoromethoxy acetophenone98
2777176
1-[2-(trifluoromethoxy)phenyl]ethanone
CC(=O)C1=CC=CC=C1OC(F)(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Density 1.265
Refractive Index 1.45
Quantity 1 g
Beilstein 2259035
Formula Weight 204.15
Percent Purity ≥98%
Chemical Name or Material 2'-(Trifluoromethoxy)acetophenone