missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethanol, 99+%, Thermo Scientific Chemicals

Catalog Number p-3779682
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
636-72-6
C5H6OS
114.16
MFCD00005454
ZPHGMBGIFODUMF-UHFFFAOYSA-N
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
69467
OCC1=CC=CS1
636-72-6
C5H6OS
114.16
MFCD00005454
ZPHGMBGIFODUMF-UHFFFAOYSA-N
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
69467
OCC1=CC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 206.0°C to 207.0°C (761.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5622 to 1.5642
Quantity 25 g
Beilstein 17, 113
Specific Gravity 0.9
Solubility Information Solubility in water: 40g/L (20°C).
IUPAC Name (thiophen-2-yl)methanol
Formula Weight 114.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiophenemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'