Learn More
2-Sulfobenzoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-Sulfobenzoic Acid
Specifications
Specifications
| Chemical Name or Material | 2-Sulfobenzoic Acid |
| CAS | 632-25-7 |
| Analyte or Component Names | 2-Sulfobenzoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H6 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzoic acid, o-sulfo- (6CI,7CI,8CI), 2-Sulfobenzoic acid, 2-Carboxybenzenesulfonic acid, o-Carboxybenzenesulfonic acid, o-Sulfobenzoic acid |
| InChI Formula | InChI=1S/C7H6O5S/c8-7(9)5-3-1-2-4-6(5)13(10,11)12/h1-4H,(H,8,9)(H,10,11,12) |
| SMILES | OC(=O)c1ccccc1S(=O)(=O)O |
| IUPAC Name | 2-sulfobenzoic acid |
| Show More |