missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Pyrrolidin-1-ylphenyl)methanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 177.25 |
Percent Purity | 95% |
Chemical Name or Material | (2-Pyrrolidin-1-ylphenyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'