Learn More
2-Propanol, Honeywell™
Catalog Number p-7145732
Click to view available options
Quantity:
2.5 L
5 L
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
Semiconductor Grade VLSI (Honeywell 17604)
  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Quantity 2.5 L
Flash Point 12 °C
Packaging Plastic Bottle
Physical Form Liquid
Refractive Index 1.3770 - 1.3780
Water Max. 500 ppm
Percent Purity min. 99.8 %
Grade Semiconductor