missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-7077194
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
127-66-2
C10H10O
146.19
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O
This item is not returnable. View return policy
127-66-2
C10H10O
146.19
KSLSOBUAIFEGLT-UHFFFAOYSA-N
2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
92098
2-phenylbut-3-yn-2-ol
CC(C#C)(C1=CC=CC=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 50.0°C to 52.0°C
Boiling Point 102.0°C to 103.0°C (12.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CC(C6H5)(OH)CH3
Quantity 5 g
Beilstein 06,II,559
Merck Index 10,7158
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone, benzene and most other common, organic solvents
Formula Weight 146.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-3-butyn-2-ol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenyl-3-butyn-2-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'