Learn More
2-Oxo Clopidogrel-13C,d3 Hydrochloride(Mixture of Diastereomers), TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | 2-Oxo Clopidogrel-13C,d3 Hydrochloride(Mixture of Diastereomers) |
| CAS | 1794713-84-0 |
| Molecular Formula | C1513CH14D3Cl2NO3S |
| Synonym | alpha-(2-Chlorophenyl)-2,6,7,7a-tetrahydro-2-oxo-thieno[3,2-c]pyridine-5(4H)-acetic Acid Methyl Ester-13C,d3 Hydrochloride; |
| InChI Formula | InChI=1S/C16H16ClNO3S.ClH/c1-21-16(20)15(11-4-2-3-5-12(11)17)18-7-6-13-10(9-18)8-14(19)22-13;/h2-5,8,13,15H,6-7,9H2,1H3;1H/i1+1D3; |
| SMILES | Cl.[2H][13C]([2H])([2H])OC(=O)C(N1CCC2SC(=O)C=C2C1)c3ccccc3Cl |
| IUPAC Name | trideuterio(1^{13}C)methyl 2-(2-chlorophenyl)-2-(2-oxo-4,6,7,7a-tetrahydrothieno[3,2-c]pyridin-5-yl)acetate;hydrochloride |
| Molecular Weight (g/mol) | 378.29 |
| Quantity | 0.5 mg |
| Formula Weight | 377.0528 g/mol |
| Show More |