Learn More
2-Nitrophenylacetonitrile, 98%, Thermo Scientific Chemicals
Catalog Number p-3779316
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
610-66-2
C8H6N2O2
162.15
MFCD00007183
YPRFCQAWSNWRLM-UHFFFAOYSA-N
2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl
11888
2-(2-nitrophenyl)acetonitrile
[O-][N+](=O)C1=CC=CC=C1CC#N
610-66-2
C8H6N2O2
162.15
MFCD00007183
YPRFCQAWSNWRLM-UHFFFAOYSA-N
2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl
11888
2-(2-nitrophenyl)acetonitrile
[O-][N+](=O)C1=CC=CC=C1CC#N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 81.0°C to 85.0°C
Color Brown to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2CN
Quantity 10 g
Beilstein 09, 455
Formula Weight 162.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Nitrophenylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrophenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.